Assay Details
| Gene Symbol: Arhgap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021493.2 | NP_067468.2 | 15 - 16 | 2988 | 106 | |
| GenBank mRNA | AK173187.1 | - | 6 - 7 | 634 | 106 | |
More Information
Set Membership: |
|

Arhgap23
Genomic Map