Assay Details
| Gene Symbol: Krtap3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023511.1 | NP_076000.1 | 1 | 219 | 84 | |
| GenBank mRNA | BC060278.1 | - | 1 | 219 | 84 | |
| D86424.1 | - | 1 | 167 | 84 | ||
More Information
Set Membership: |
|

Krtap3-1
Genomic Map