Assay Details
| Gene Symbol: Glt6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039095.1 | NP_001034184.1 | 3 - 4 | 194 | 80 | |
| GenBank mRNA | BC111823.1 | - | 3 - 4 | 194 | 40052059 | 80 |
More Information
Set Membership: |
|

Glt6d1
Genomic Map