Assay Details
| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026114.3 | NP_080390.1 | 1 - 2 | 111 | 85 | |
| GenBank mRNA | AK003094.1 | - | 1 - 2 | 418 | 85 | |
| AK010592.1 | - | 1 - 2 | 141 | 85 | ||
| AK143499.1 | - | 1 - 2 | 112 | 85 | ||
| AK167763.1 | - | 1 - 2 | 113 | 85 | ||
| BC005463.1 | - | 1 - 2 | 91 | 3591411 | 85 | |
| BC016448.1 | - | 1 - 2 | 115 | 4234809 | 85 | |
| BC016497.1 | - | 1 - 2 | 115 | 4501321 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010592.1, AK143499.1, AK167763.1, BC005463.1, BC016448.1, BC016497.1, NM_026114.3 transcripts.
Set Membership: |
|

Eif2s1
Genomic Map