Assay Details
| Gene Symbol: Rab32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026405.3 | NP_080681.1 | 2 - 3 | 637 | 78 | |
| GenBank mRNA | AB232627.1 | - | 2 - 3 | 518 | 78 | |
| AK012701.1 | - | 2 - 3 | 649 | 78 | ||
| AK078874.1 | - | 2 - 3 | 329 | 78 | ||
| AK088661.1 | - | 2 - 3 | 638 | 78 | ||
| AK151543.1 | - | 2 - 3 | 611 | 78 | ||
| AK159102.1 | - | 2 - 3 | 664 | 78 | ||
| AK159901.1 | - | 2 - 3 | 664 | 78 | ||
| AK163620.1 | - | 2 - 3 | 588 | 78 | ||
| AK206948.1 | - | 2 - 3 | 347 | 78 | ||
| BC016409.1 | - | 555 | 4456902 | 78 | ||
| BC055945.1 | - | 563 | 4161169 | 78 | ||
More Information
Set Membership: |
|

Rab32
Genomic Map