Assay Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026695.3 | NP_080971.2 | 4 - 5 | 514 | 68 | |
| GenBank mRNA | AK002407.1 | - | 4 - 5 | 456 | 68 | |
| AK013284.1 | - | 4 - 5 | 478 | 68 | ||
| AK150293.1 | - | 4 - 5 | 457 | 68 | ||
| BC049237.1 | - | 4 - 5 | 443 | 4241552 | 68 | |
| BC069877.1 | - | 4 - 5 | 473 | 5370580 | 68 | |
More Information
Set Membership: |
|

Etfb
Genomic Map