Assay Details
| Gene Symbol: Snrpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026943.1 | NP_081219.1 | 2 - 3 | 237 | 81 | |
| GenBank mRNA | AK007389.1 | - | 2 - 3 | 216 | 81 | |
| AK045968.1 | - | 2 - 3 | 212 | 81 | ||
| AK088105.1 | - | 2 - 3 | 216 | 81 | ||
| BC043014.1 | - | 2 - 3 | 237 | 5683364 | 81 | |
| BC051208.1 | - | 1 - 2 | 216 | 1510565 | 81 | |
| BC116858.1 | - | 2 - 3 | 220 | 40126177 | 81 | |
| BC116860.1 | - | 2 - 3 | 220 | 40126179 | 81 | |
More Information
Set Membership: |
|

Snrpd2
Genomic Map