Assay Details
| Gene Symbol: Gemin7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027189.2 | NP_081465.1 | 1 - 2 | 42 | 60 | |
| GenBank mRNA | AK010303.1 | - | 1 - 2 | 44 | 60 | |
| BC028527.1 | - | 1 - 2 | 38 | 1362579 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010303.1, BC028527.1, NM_027189.2 transcripts.
Set Membership: |
|

Gemin7
Genomic Map