Assay Details
| Gene Symbol: Gpr39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027677.2 | NP_081953.2 | 1 - 2 | 1195 | 106 | |
| GenBank mRNA | BC085285.1 | - | 1 - 2 | 1208 | 30745457 | 106 |
More Information
Set Membership: |
|

Gpr39
Genomic Map