Assay Details
| Gene Symbol: Ercc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028042.3 | NP_082318.2 | 8 - 9 | 777 | 87 | |
| GenBank mRNA | AJ427222.1 | - | 8 - 9 | 721 | 87 | |
| AK010576.1 | - | 8 - 9 | 779 | 87 | ||
| AK014517.1 | - | 7 - 8 | 677 | 87 | ||
| AK082109.1 | - | 7 - 8 | 667 | 87 | ||
| AK156267.1 | - | 8 - 9 | 765 | 87 | ||
| AK157175.1 | - | 8 - 9 | 778 | 87 | ||
| AK157504.1 | - | 7 - 8 | 676 | 87 | ||
| BC037200.1 | - | 8 - 9 | 757 | 3985548 | 87 | |
| BC056463.1 | - | 8 - 9 | 761 | 87 | ||
More Information
Set Membership: |
|

Ercc8
Genomic Map