Assay Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028462.1 | NP_082738.1 | 2 - 3 | 1128 | 95 | |
| GenBank mRNA | AK014242.1 | - | 2 - 3 | 1006 | 95 | |
| BC049157.1 | - | 2 - 3 | 1227 | 95 | ||
| BC119565.1 | - | 2 - 3 | 1128 | 40057718 | 95 | |
| BC119566.1 | - | 2 - 3 | 1128 | 40057721 | 95 | |
More Information
Set Membership: |
|

Fezf1
Genomic Map