Assay Details
| Gene Symbol: Rai14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166408.1 | NP_001159880.1 | 3 - 4 | 328 | 109 | |
| NM_030690.3 | NP_109615.1 | 3 - 4 | 414 | 109 | ||
| GenBank mRNA | AF202315.1 | - | 2 - 3 | 201 | 109 | |
| AF274866.1 | - | 3 - 4 | 393 | 109 | ||
| AK090343.1 | - | 3 - 4 | 323 | 109 | ||
| AK129333.1 | - | 3 - 4 | 264 | 109 | ||
| AK160119.1 | - | 3 - 4 | 324 | 109 | ||
| BC052458.1 | - | 3 - 4 | 414 | 6830677 | 109 | |
More Information
Set Membership: |
|

Rai14
Genomic Map