Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 12 - 13 | 2974 | 87 | |
| GenBank mRNA | AF330197.1 | - | 12 - 13 | 2970 | 87 | |
| AK036351.1 | - | 11 - 12 | 2709 | 87 | ||
| AK048110.1 | - | 11 - 12 | 2740 | 87 | ||
| AK137148.1 | - | 10 - 11 | 2312 | 87 | ||
| AK164502.1 | - | 3 - 4 | 371 | 87 | ||
| AK169999.1 | - | 11 - 12 | 3080 | 87 | ||
| BC011463.1 | - | 6 - 7 | 1406 | 87 | ||
| BC026485.1 | - | 3 - 4 | 335 | 87 | ||
| BC054842.1 | - | 2 - 3 | 213 | 87 | ||
| BC069835.1 | - | 12 - 13 | 2830 | 30094289 | 87 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map