Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 5 - 6 | 1507 | 92 | |
| GenBank mRNA | AF330197.1 | - | 5 - 6 | 1503 | 92 | |
| AK036351.1 | - | 4 - 5 | 1242 | 92 | ||
| AK048110.1 | - | 4 - 5 | 1273 | 92 | ||
| AK137148.1 | - | 3 - 4 | 845 | 92 | ||
| AK169999.1 | - | 4 - 5 | 1617 | 92 | ||
| AK171932.1 | - | 4 - 5 | 1445 | 92 | ||
| BC069835.1 | - | 5 - 6 | 1363 | 30094289 | 92 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map