Assay Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033604.2 | NP_291082.1 | 6 - 7 | 1715 | 66 | |
| GenBank mRNA | AF330197.1 | - | 6 - 7 | 1711 | 66 | |
| AK036351.1 | - | 5 - 6 | 1450 | 66 | ||
| AK048110.1 | - | 5 - 6 | 1481 | 66 | ||
| AK137148.1 | - | 4 - 5 | 1053 | 66 | ||
| AK169999.1 | - | 5 - 6 | 1825 | 66 | ||
| BC069835.1 | - | 6 - 7 | 1571 | 30094289 | 66 | |
More Information
Set Membership: |
|

Rnf111
Genomic Map