Assay Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138667.3 | NP_619608.1 | 2 - 3 | 502 | 114 | |
| GenBank mRNA | AB093262.1 | - | 1 - 2 | 69 | 114 | |
| AK147830.1 | - | 2 - 3 | 313 | 114 | ||
| AY093701.1 | - | 1 - 2 | 162 | 114 | ||
| BC004813.1 | - | 1 - 2 | 169 | 3586132 | 114 | |
| CK391470.1 | - | 3 - 4 | 368 | 114 | ||
More Information
Set Membership: |
|

Tab2
Genomic Map