Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 9 - 10 | 972 | 75 | |
| NM_001164243.1 | NP_001157715.1 | 9 - 10 | 1015 | 75 | ||
| NM_001164244.1 | NP_001157716.1 | 8 - 9 | 919 | 75 | ||
| NM_144806.2 | NP_659055.1 | 10 - 11 | 1068 | 75 | ||
| GenBank mRNA | AK038680.1 | - | 10 - 11 | 1034 | 75 | |
| AK076811.1 | - | 10 - 11 | 1022 | 75 | ||
| AK077330.1 | - | 10 - 11 | 1021 | 75 | ||
| BC023178.1 | - | 10 - 11 | 1053 | 4947226 | 75 | |
| BC051438.1 | - | 9 - 10 | 939 | 6311509 | 75 | |
| BC106185.1 | - | 9 - 10 | 901 | 6512229 | 75 | |
More Information
Set Membership: |
|

Prpsap2
Genomic Map