Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 10 - 11 | 1116 | 80 | |
| NM_001164243.1 | NP_001157715.1 | 10 - 11 | 1159 | 80 | ||
| NM_001164244.1 | NP_001157716.1 | 9 - 10 | 1063 | 80 | ||
| NM_144806.2 | NP_659055.1 | 11 - 12 | 1212 | 80 | ||
| GenBank mRNA | AK038680.1 | - | 11 - 12 | 1178 | 80 | |
| AK076811.1 | - | 11 - 12 | 1166 | 80 | ||
| AK077330.1 | - | 11 - 12 | 1165 | 80 | ||
| BC023178.1 | - | 11 - 12 | 1197 | 4947226 | 80 | |
| BC051438.1 | - | 10 - 11 | 1083 | 6311509 | 80 | |
| BC106185.1 | - | 10 - 11 | 1045 | 6512229 | 80 | |
More Information
Set Membership: |
|

Prpsap2
Genomic Map