Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164243.1 | NP_001157715.1 | 2 - 3 | 225 | 76 | |
| NM_144806.2 | NP_659055.1 | 2 - 3 | 225 | 76 | ||
| GenBank mRNA | AK038680.1 | - | 2 - 3 | 191 | 76 | |
| AK076811.1 | - | 2 - 3 | 179 | 76 | ||
| AK077330.1 | - | 2 - 3 | 178 | 76 | ||
| AK205638.1 | - | 2 - 3 | 110 | 76 | ||
| BC023178.1 | - | 2 - 3 | 210 | 4947226 | 76 | |
| BC051438.1 | - | 2 - 3 | 149 | 6311509 | 76 | |
| BY204265.1 | - | 2 - 3 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038680.1, AK076811.1, AK077330.1, BC023178.1, BC051438.1, NM_001164243.1, NM_144806.2 transcripts.
Set Membership: |
|

Prpsap2
Genomic Map