Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 3 - 4 | 331 | 66 | |
| NM_144806.2 | NP_659055.1 | 4 - 5 | 427 | 66 | ||
| GenBank mRNA | AK038680.1 | - | 4 - 5 | 393 | 66 | |
| AK076811.1 | - | 4 - 5 | 381 | 66 | ||
| AK077330.1 | - | 4 - 5 | 380 | 66 | ||
| BC023178.1 | - | 4 - 5 | 412 | 4947226 | 66 | |
| BC106185.1 | - | 3 - 4 | 260 | 6512229 | 66 | |
More Information
Set Membership: |
|

Prpsap2
Genomic Map