Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 4 - 5 | 412 | 93 | |
| NM_001164243.1 | NP_001157715.1 | 4 - 5 | 455 | 93 | ||
| NM_001164244.1 | NP_001157716.1 | 3 - 4 | 359 | 93 | ||
| NM_144806.2 | NP_659055.1 | 5 - 6 | 508 | 93 | ||
| GenBank mRNA | AK038680.1 | - | 5 - 6 | 474 | 93 | |
| AK076811.1 | - | 5 - 6 | 462 | 93 | ||
| AK077330.1 | - | 5 - 6 | 461 | 93 | ||
| BC023178.1 | - | 5 - 6 | 493 | 4947226 | 93 | |
| BC051438.1 | - | 4 - 5 | 379 | 6311509 | 93 | |
| BC106185.1 | - | 4 - 5 | 341 | 6512229 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051438.1, NM_001164243.1, NM_001164244.1 transcripts.
Set Membership: |
|

Prpsap2
Genomic Map