Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 5 - 6 | 581 | 90 | |
| NM_001164243.1 | NP_001157715.1 | 5 - 6 | 624 | 90 | ||
| NM_001164244.1 | NP_001157716.1 | 4 - 5 | 528 | 90 | ||
| NM_144806.2 | NP_659055.1 | 6 - 7 | 677 | 90 | ||
| GenBank mRNA | AK038680.1 | - | 6 - 7 | 643 | 90 | |
| AK076811.1 | - | 6 - 7 | 631 | 90 | ||
| AK077330.1 | - | 6 - 7 | 630 | 90 | ||
| BC023178.1 | - | 6 - 7 | 662 | 4947226 | 90 | |
| BC051438.1 | - | 5 - 6 | 548 | 6311509 | 90 | |
| BC106185.1 | - | 5 - 6 | 510 | 6512229 | 90 | |
More Information
Set Membership: |
|

Prpsap2
Genomic Map