Assay Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1221 | 107 | |
| NR_073436.1 | 1 | 1221 | 107 | |||
| NR_073437.1 | 1 | 1221 | 107 | |||
| NR_073438.1 | 1 | 1221 | 107 | |||
| GenBank mRNA | AA028814.1 | - | 4 | 401 | 107 | |
| AF408398.1 | - | 1 | 893 | 107 | ||
| BG094109.1 | - | 1 | 333 | 107 | ||
| BI990274.1 | - | 1 | 160 | 107 | ||
| CX237465.1 | - | 1 | 273 | 107 | ||
More Information
Set Membership: |
|

Crebzf
Genomic Map