Assay Details
| Gene Symbol: March2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145486.5 | NP_663461.2 | 5 - 6 | 903 | 71 | |
| NR_045522.1 | 5 - 6 | 903 | 71 | |||
| GenBank mRNA | AK039778.1 | - | 5 - 6 | 896 | 71 | |
| AK053434.1 | - | 5 - 6 | 871 | 71 | ||
| BC132541.1 | - | 4 - 5 | 779 | 40130818 | 71 | |
| BC138064.1 | - | 4 - 5 | 779 | 8861083 | 71 | |
More Information
Set Membership: |
|

March2
Genomic Map