Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145552.2 | NP_663527.2 | 11 - 12 | 1422 | 68 | |
| GenBank mRNA | AK037796.1 | - | 11 - 12 | 1358 | 68 | |
| AK077532.1 | - | 11 - 12 | 1392 | 68 | ||
| BC003262.1 | - | 11 - 12 | 1398 | 3501393 | 68 | |
| U69600.1 | - | 11 - 12 | 1317 | 68 | ||
More Information
Set Membership: |
|

Gnl2
Genomic Map