Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145552.2 | NP_663527.2 | 5 - 6 | 687 | 89 | |
| GenBank mRNA | AK037796.1 | - | 5 - 6 | 623 | 89 | |
| AK077532.1 | - | 5 - 6 | 657 | 89 | ||
| AK146404.1 | - | 5 - 6 | 684 | 89 | ||
| BC003262.1 | - | 5 - 6 | 663 | 3501393 | 89 | |
More Information
Set Membership: |
|

Gnl2
Genomic Map