Assay Details
| Gene Symbol: Irf2bpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145836.2 | NP_665835.1 | 1 | 2221 | 71 | |
| GenBank mRNA | AF525300.1 | - | 1 | 2218 | 71 | |
| AK032366.1 | - | 1 | 2226 | 71 | ||
| AK082791.1 | - | 1 | 2205 | 71 | ||
| AK084782.1 | - | 1 | 2229 | 71 | ||
| AK140441.1 | - | 1 | 2202 | 71 | ||
| AK154383.1 | - | 1 | 1971 | 71 | ||
| AK155373.1 | - | 1 | 2176 | 71 | ||
| AK160059.1 | - | 1 | 2175 | 71 | ||
| AK171043.1 | - | 1 | 2176 | 71 | ||
| AK173282.1 | - | 1 | 2221 | 71 | ||
| BC049246.1 | - | 2 | 424 | 71 | ||
| BC057128.1 | - | 1 | 1658 | 6810092 | 71 | |
| BC063253.1 | - | 1 | 1658 | 6810092 | 71 | |
More Information
Set Membership: |
|

Irf2bpl
Genomic Map