Assay Details
| Gene Symbol: Cables2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145851.2 | NP_665850.2 | 9 - 10 | 1301 | 85 | |
| GenBank mRNA | AK050121.1 | - | 6 - 7 | 2923 | 85 | |
| BC024565.1 | - | 2 - 3 | 312 | 85 | ||
| BC036954.1 | - | 4 - 5 | 692 | 85 | ||
| BC037028.1 | - | 4 - 5 | 692 | 85 | ||
| BC052789.1 | - | 3 - 4 | 643 | 85 | ||
More Information
Set Membership: |
|

Cables2
Genomic Map