Assay Details
| Gene Symbol: Purg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098233.1 | NP_001091703.1 | 1 | 720 | 70 | |
| NM_152821.2 | NP_690034.1 | 1 | 720 | 70 | ||
| GenBank mRNA | AF479672.1 | - | 1 | 720 | 70 | |
| AF479673.1 | - | 1 | 720 | 70 | ||
| AK015630.1 | - | 2 | 731 | 70 | ||
| BC116406.2 | - | 1 | 720 | 40102393 | 70 | |
| CD772246.1 | - | 1 | 285 | 70 | ||
More Information
Set Membership: |
|

Purg
Genomic Map