Assay Details
| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167976.1 | NP_001161448.1 | 3 - 4 | 207 | 96 | |
| NM_172577.3 | NP_766165.2 | 2 - 3 | 650 | 96 | ||
| GenBank mRNA | AK036988.1 | - | 2 - 3 | 390 | 96 | |
| AK041694.1 | - | 2 - 3 | 651 | 96 | ||
| AK044945.1 | - | 2 - 3 | 153 | 96 | ||
| AK045180.1 | - | 2 - 3 | 476 | 96 | ||
| AK167535.1 | - | 2 - 3 | 358 | 96 | ||
| BC057980.1 | - | 2 - 3 | 474 | 5042653 | 96 | |
| EU180236.1 | - | 3 - 4 | 207 | 96 | ||
More Information
Set Membership: |
|

Slc25a21
Genomic Map