Assay Details
| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167976.1 | NP_001161448.1 | 8 - 9 | 688 | 93 | |
| NM_172577.3 | NP_766165.2 | 7 - 8 | 1131 | 93 | ||
| GenBank mRNA | AK036988.1 | - | 7 - 8 | 871 | 93 | |
| AK041694.1 | - | 7 - 8 | 1132 | 93 | ||
| AK045180.1 | - | 7 - 8 | 957 | 93 | ||
| AK162379.1 | - | 7 - 8 | 958 | 93 | ||
| AK167535.1 | - | 7 - 8 | 839 | 93 | ||
| BC057980.1 | - | 7 - 8 | 955 | 5042653 | 93 | |
| EU180236.1 | - | 8 - 9 | 688 | 93 | ||
More Information
Set Membership: |
|

Slc25a21
Genomic Map