Assay Details
| Gene Symbol: Arsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173451.3 | NP_775627.1 | 1 - 2 | 1308 | 87 | |
| GenBank mRNA | AK034454.1 | - | 1 - 2 | 1309 | 87 | |
| AK034584.1 | - | 1 - 2 | 1332 | 87 | ||
| AK046410.1 | - | 1 - 2 | 1292 | 87 | ||
| AK052931.1 | - | 1 - 2 | 1289 | 87 | ||
| BC117813.2 | - | 1 - 2 | 528 | 40092671 | 87 | |
More Information
Set Membership: |
|

Arsj
Genomic Map