Assay Details
| Gene Symbol: Rab39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175562.3 | NP_780771.1 | 1 - 2 | 341 | 85 | |
| GenBank mRNA | AB232635.1 | - | 1 - 2 | 224 | 85 | |
| AK049038.1 | - | 1 - 2 | 341 | 85 | ||
| AK053487.1 | - | 1 - 2 | 338 | 85 | ||
| BC049787.1 | - | 1 - 2 | 321 | 6511404 | 85 | |
More Information
Set Membership: |
|

Rab39
Genomic Map