Assay Details
| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177233.5 | NP_796207.2 | 2 - 3 | 416 | 104 | |
| GenBank mRNA | AK081531.1 | - | 2 - 3 | 399 | 104 | |
| AK149355.1 | - | 2 - 3 | 419 | 104 | ||
| AY325123.1 | - | 2 - 3 | 199 | 104 | ||
| BB642822.1 | - | 2 - 3 | 416 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK081531.1, AK149355.1, AY325123.1, NM_177233.5 transcripts.
Set Membership: |
|

Fam19a4
Genomic Map