Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177473.3 | NP_803424.2 | 2 - 3 | 298 | 60 | |
| GenBank mRNA | BU936413.1 | - | 2 - 3 | 281 | 60 | |
| BY296351.1 | - | 2 - 3 | 153 | 60 | ||
More Information
Set Membership: |
|

Tmem191c
Genomic Map