Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177473.3 | NP_803424.2 | 6 - 7 | 546 | 72 | |
| GenBank mRNA | BC058760.1 | - | 2 - 3 | 641 | 72 | |
| BC098238.1 | - | 2 - 3 | 173 | 72 | ||
| BU936413.1 | - | 6 - 7 | 529 | 72 | ||
More Information
Set Membership: |
|

Tmem191c
Genomic Map