Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177473.3 | NP_803424.2 | 7 - 8 | 605 | 94 | |
| GenBank mRNA | AB041601.1 | - | 4 - 5 | 922 | 94 | |
| BC058760.1 | - | 3 - 4 | 700 | 94 | ||
| BC098238.1 | - | 3 - 4 | 232 | 94 | ||
More Information
Set Membership: |
|

Tmem191c
Genomic Map