Assay Details
| Gene Symbol: Tmem191c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177473.3 | NP_803424.2 | 9 - 10 | 724 | 100 | |
| GenBank mRNA | AB041601.1 | - | 6 - 7 | 1041 | 100 | |
| BC058760.1 | - | 5 - 6 | 819 | 100 | ||
| BC098238.1 | - | 5 - 6 | 351 | 100 | ||
More Information
Set Membership: |
|

Tmem191c
Genomic Map