Assay Details
| Gene Symbol: Fam83g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178618.3 | NP_848733.2 | 1 - 2 | 653 | 71 | |
| GenBank mRNA | AK028566.1 | - | 1 - 2 | 600 | 71 | |
| BC085279.1 | - | 1 - 2 | 614 | 30438273 | 71 | |
More Information
Set Membership: |
|

Fam83g
Genomic Map