Assay Details
| Gene Symbol: Defb37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181683.2 | NP_859011.1 | 1 - 2 | 146 | 86 | |
| GenBank mRNA | AJ489588.1 | - | 1 - 2 | 146 | 86 | |
| AJ578470.1 | - | 1 - 2 | 64 | 86 | ||
More Information
Set Membership: |
|

Defb37
Genomic Map