Assay Details
| Gene Symbol: Fam24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183272.2 | NP_899095.1 | 1 - 2 | 35 | 68 | |
| GenBank mRNA | AK006815.1 | - | 1 - 2 | 36 | 68 | |
| AK018853.1 | - | 1 - 2 | 37 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006815.1, NM_183272.2 transcripts.
Set Membership: |
|

Fam24a
Genomic Map