Assay Details
| Gene Symbol: Tigd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207278.2 | NP_997161.1 | 1 | 1422 | 93 | |
| GenBank mRNA | AK081667.1 | - | 1 | 1424 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tigd4| Gene Symbol: Tigd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207278.2 | NP_997161.1 | 1 | 1422 | 93 | |
| GenBank mRNA | AK081667.1 | - | 1 | 1424 | 93 | |
Set Membership: |
|