Assay Details
| Gene Symbol: Pwwp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098636.1 | NP_001092106.1 | 2 - 3 | 1855 | 70 | |
| GenBank mRNA | AK213732.1 | - | 1 - 2 | 324 | 70 | |
More Information
Set Membership: |
|

Pwwp2b
Genomic Map