Assay Details
| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109749.1 | NP_001103219.1 | 16 - 17 | 2158 | 93 | |
| NM_001109751.1 | NP_001103221.1 | 15 - 16 | 2246 | 93 | ||
| NM_173004.3 | NP_766592.2 | 16 - 17 | 2158 | 93 | ||
| GenBank mRNA | AK036262.1 | - | 16 - 17 | 2128 | 93 | |
| AK173340.1 | - | 9 - 10 | 1277 | 93 | ||
| BC046975.1 | - | 13 - 14 | 1780 | 93 | ||
| BC115766.1 | - | 16 - 17 | 2048 | 40060288 | 93 | |
More Information
Set Membership: |
|

Cntn4
Genomic Map