Assay Details
| Gene Symbol: Mgat4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145300.2 | NP_001138772.1 | 2 - 3 | 510 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4e| Gene Symbol: Mgat4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145300.2 | NP_001138772.1 | 2 - 3 | 510 | 76 | |
Set Membership: |
|