Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160164.1 | NP_001153636.1 | 1 - 2 | 105 | 94 | |
| GenBank mRNA | AK145284.1 | - | 1 - 2 | 106 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145284.1, NM_001160164.1 transcripts.
Set Membership: |
|

Neu2
Genomic Map