Assay Details
| Gene Symbol: Prpsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164242.1 | NP_001157714.1 | 1 - 2 | 133 | 78 | |
| NM_001164244.1 | NP_001157716.1 | 1 - 2 | 133 | 78 | ||
| GenBank mRNA | BC106185.1 | - | 1 - 2 | 62 | 6512229 | 78 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC106185.1, NM_001164242.1, NM_001164244.1 transcripts.
Set Membership: |
|

Prpsap2
Genomic Map