Assay Details
| Gene Symbol: Kir3dl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177749.3 | NP_808417.2 | 6 - 7 | 1016 | 107 | |
| GenBank mRNA | AY130461.1 | - | 6 - 7 | 1002 | 107 | |
| AY152727.1 | - | 5 - 6 | 737 | 107 | ||
| AY530782.1 | - | 6 - 7 | 989 | 107 | ||
| AY530783.1 | - | 6 - 7 | 989 | 107 | ||
| BC141012.1 | - | 6 - 7 | 1002 | 9055624 | 107 | |
More Information
Set Membership: |
|

Kir3dl1
Genomic Map