Assay Details
| Gene Symbol: Prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF004670.1 | - | 2 - 3 | 269 | 111 | |
| AF093852.1 | - | 2 - 3 | 300 | 111 | ||
| AK030413.1 | - | 2 - 3 | 302 | 111 | ||
| AK157820.1 | - | 2 - 3 | 319 | 111 | ||
| AK161710.1 | - | 2 - 3 | 302 | 111 | ||
| AK168223.1 | - | 2 - 3 | 273 | 111 | ||
| AK172888.1 | - | 2 - 3 | 262 | 111 | ||
| BC013489.1 | - | 2 - 3 | 261 | 4215591 | 111 | |
| BC061181.1 | - | 2 - 3 | 301 | 30309446 | 111 | |
| Y12883.1 | - | 2 - 3 | 319 | 111 | ||
More Information
Set Membership: |
|

Prdx6
Genomic Map