Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009214.3 | NP_033240.3 | 1 - 2 | 144 | 90 | |
| GenBank mRNA | AF031486.1 | - | 1 - 2 | 127 | 90 | |
| AK002687.1 | - | 1 - 2 | 142 | 90 | ||
| AK011542.1 | - | 1 - 2 | 146 | 90 | ||
| AK017775.1 | - | 1 - 2 | 115 | 90 | ||
| AK049780.1 | - | 1 - 2 | 143 | 90 | ||
| AK049787.1 | - | 1 - 2 | 133 | 90 | ||
| AK136798.1 | - | 1 - 2 | 145 | 90 | ||
| AK149929.1 | - | 1 - 2 | 142 | 90 | ||
| AK160637.1 | - | 1 - 2 | 139 | 90 | ||
| AK160659.1 | - | 1 - 2 | 145 | 90 | ||
| AK166665.1 | - | 1 - 2 | 170 | 90 | ||
| BC046623.1 | - | 1 - 2 | 126 | 6315420 | 90 | |
| BC058688.1 | - | 1 - 2 | 385 | 6413869 | 90 | |
| Y09419.1 | - | 1 - 2 | 117 | 90 | ||
More Information
Set Membership: |
|

Sms
Genomic Map